1-Fluoro-2,4-dimethyl-5-nitrobenzene ≥95%

Supplier: Ambeed
Warning

A167320-1G A167320-250MG A167320-5G
77885-320EA 120 USD
77885-320 77885-321 77885-322
1-Fluoro-2,4-dimethyl-5-nitrobenzene ≥95%
1-Fluoro-2,4-dimethyl-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₈H₈FNO₂
MDL Number: MFCD12545823
CAS Number: 345-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR