4-Chloro-2-(1-((2,3-dimethylphenyl)amino)ethyl)phenol ≥98%

Supplier: Ambeed
Warning

Synonyms: MFI8

A1671394-10MG A1671394-1MG A1671394-5MG
77598-150EA 78.93 USD
77598-150 77598-152 77598-154
4-Chloro-2-(1-((2,3-dimethylphenyl)amino)ethyl)phenol ≥98%
4-Chloro-2-(1-((2,3-dimethylphenyl)amino)ethyl)phenol

Precaución: for Research Use Only

Formula: C₁₆H₁₈ClNO
MW: 275.77 g/mol
CAS Number: 694488-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR