(E)-2-(1-(5-Chloro-2-hydroxyphenyl)ethylidene)-N-phenylhydrazine-1-carbothioamide ≥95%

Supplier: Ambeed
Danger

A1671344-100MG A1671344-250MG
77876-537EA 235 USD
77876-537 77876-538
(E)-2-(1-(5-Chloro-2-hydroxyphenyl)ethylidene)-N-phenylhydrazine-1-carbothioamide ≥95%
(E)-2-(1-(5-Chloro-2-hydroxyphenyl)ethylidene)-N-phenylhydrazine-1-carbothioamide

Precaución: for Research Use Only

Formula: C₁₅H₁₄ClN₃OS
CAS Number: 2426686-17-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR