Pyridine-2,6-diyldimethanamine ≥98%

Supplier: Ambeed
Danger

A167080-250MG A167080-5G A167080-100MG
77593-018EA 32.8 USD
77593-018 77748-107 77748-106
Pyridine-2,6-diyldimethanamine ≥98%
Pyridine-2,6-diyldimethanamine

Precaución: for Research Use Only

Formula: C₇H₁₁N₃
MW: 137.1823 g/mol
MDL Number: MFCD06213715
CAS Number: 34984-16-2
UN: 2735
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR