6-Azaindole-2-carboxylic acid ≥90%

Supplier: Ambeed
Warning

Synonyms: 1H-Pyrrolo[2,3-c]pyridine-2-carboxylic acid, 2-Carboxy-1H-pyrrolo[2,3-c]pyridine

A167072-250MG A167072-100MG A167072-1G A167072-5G
77611-628EA 37.08 USD
77611-628 77671-358 77671-360 77881-291
6-Azaindole-2-carboxylic acid ≥90%
6-Azaindole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MDL Number: MFCD11506121
CAS Number: 24334-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR