trans-1,4-Bis(bromomethyl)cyclohexane ≥95%

Supplier: Ambeed
Warning

A1670291-1G A1670291-5G
77876-523EA 178 USD
77876-523 77876-524
trans-1,4-Bis(bromomethyl)cyclohexane ≥95%
trans-1,4-Bis(bromomethyl)cyclohexane

Precaución: for Research Use Only

Formula: C₈H₁₄Br₂
MW: 270 g/mol
CAS Number: 57702-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR