(1S,3R,4S,5R)-3,4,5-Tris(((E)-3-(3,4-dihydroxyphenyl)acryloyl)oxy)-1-hydroxycyclohexane-1-carboxylic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: 3,4,5-Tricaffeoylquinic acid

A1669944-10MG A1669944-25MG A1669944-5MG A1669944-1MG
77598-134EA 785.15 USD
77598-134 77598-136 77598-138 77625-650
(1S,3R,4S,5R)-3,4,5-Tris(((E)-3-(3,4-dihydroxyphenyl)acryloyl)oxy)-1-hydroxycyclohexane-1-carboxylic acid ≥98%
(1S,3R,4S,5R)-3,4,5-Tris(((E)-3-(3,4-dihydroxyphenyl)acryloyl)oxy)-1-hydroxycyclohexane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₃₄H₃₀O₁₅
MDL Number: MFCD09970462
CAS Number: 86632-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR