[2,2':6',2''-Terpyridine]-6,6''-dicarboxylic acid ≥95%

Supplier: Ambeed
Warning

A1669576-1G A1669576-250MG
77625-646EA 983.65 USD
77625-646 77625-648
[2,2':6',2''-Terpyridine]-6,6''-dicarboxylic acid ≥95%
[2,2':6',2''-Terpyridine]-6,6''-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₁₇H₁₁N₃O₄
MW: 321.29 g/mol
MDL Number: MFCD09030947
CAS Number: 149982-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR