(3-Cyanopropyl)boronic acid ≥95%

Supplier: Ambeed
Danger

A1667787-100MG A1667787-1G A1667787-250MG
77876-512EA 46 USD
77876-512 77876-513 77876-514
(3-Cyanopropyl)boronic acid ≥95%
(3-Cyanopropyl)boronic acid

Precaución: for Research Use Only

Formula: C₄H₈BNO₂
CAS Number: 1352619-22-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR