2,2'-(Octanedioylbis(oxy))bis(N,N,N-trimethylethan-1-aminium) chloride ≥98%

Supplier: Ambeed
Danger

Synonyms: Suberyl dicholine dichloride

A1667077-1G A1667077-250MG
77625-640EA 63.86 USD
77625-640 77625-642
2,2'-(Octanedioylbis(oxy))bis(N,N,N-trimethylethan-1-aminium) chloride ≥98%
2,2'-(Octanedioylbis(oxy))bis(N,N,N-trimethylethan-1-aminium) chloride

Precaución: for Research Use Only

Formula: C₁₈H₃₈Cl₂N₂O₄
MW: 417.41 g/mol
MDL Number: MFCD00011784
CAS Number: 100930-12-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR