(4-Benzylpiperazin-1-yl)(2-(isopentylamino)pyridin-3-yl)methanone 98+%

Supplier: Ambeed
Warning

Synonyms: NSI-189

A166690-100MG A166690-50MG
77685-356EA 8 USD
77685-356 77685-357
(4-Benzylpiperazin-1-yl)(2-(isopentylamino)pyridin-3-yl)methanone 98+%
(4-Benzylpiperazin-1-yl)(2-(isopentylamino)pyridin-3-yl)methanone

Precaución: for Research Use Only

Formula: C₂₂H₃₀N₄O
MW: 366.5 g/mol
MDL Number: MFCD26959792
CAS Number: 1270138-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR