2,6-Difluoro-4-iodonicotinaldehyde ≥98%
Supplier: Ambeed
|
Warning
|
A1666112-1G
A1666112-250MG
77876-507EA
22
USD
77876-507
77876-508
2,6-Difluoro-4-iodonicotinaldehyde ≥98%
2,6-Difluoro-4-iodonicotinaldehyde
Precaución: for Research Use Only
Formula:
C₆H₂F₂INO |
MDL Number:
MFCD34603408 CAS Number: 2417489-94-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...