(4-((1-(tert-Butoxycarbonyl)azetidin-3-yl)oxy)phenyl)boronic acid ≥95%

Supplier: Ambeed
Warning

A1665131-100MG A1665131-1G A1665131-250MG A1665131-5G
77876-498EA 198 USD
77876-498 77876-499 77876-500 77876-501
(4-((1-(tert-Butoxycarbonyl)azetidin-3-yl)oxy)phenyl)boronic acid ≥95%
(4-((1-(tert-Butoxycarbonyl)azetidin-3-yl)oxy)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₄H₂₀BNO₅
MDL Number: MFCD18383746
CAS Number: 1224449-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR