4-Amino-N-(5-chloro-2-methylphenyl)benzamide ≥97%

Supplier: Ambeed
Warning

A166114-1G A166114-5G
77671-332EA 75 USD
77671-332 77881-288
4-Amino-N-(5-chloro-2-methylphenyl)benzamide ≥97%
4-Amino-N-(5-chloro-2-methylphenyl)benzamide

Precaución: for Research Use Only

Formula: C₁₄H₁₃ClN₂O
MDL Number: MFCD03166474
CAS Number: 897595-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR