2-(1-Methyl-1H-imidazol-5-yl)ethanamine dihydrochloride ≥97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Methylhistamine 2HCl
A166042-100MG
A166042-10MG
A166042-5MG
A166042-1G
A166042-250MG
A166042-50MG
77667-322EA
298
USD
77667-322
77667-324
77667-326
77871-831
77871-832
77871-833
2-(1-Methyl-1H-imidazol-5-yl)ethanamine dihydrochloride ≥97%
2-(1-Methyl-1H-imidazol-5-yl)ethanamine dihydrochloride
Precaución: for Research Use Only
Formula:
C₆H₁₃Cl₂N₃ |
MDL Number:
MFCD00037002 CAS Number: 36475-47-5 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...