7-Hydroxychromone ≥97%

Supplier: Ambeed
Warning

A165424-100MG A165424-1G A165424-250MG
77883-291EA 16 USD
77883-291 77883-292 77883-293
7-Hydroxychromone ≥97%
7-Hydroxychromone

Precaución: for Research Use Only

Formula: C₉H₆O₃
MDL Number: MFCD00209371
CAS Number: 59887-89-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR