8-Amino-N-(1-(cyclopropanecarbonyl)azetidin-3-yl)-2-(3-fluorophenyl)-1,7-naphthyridine-5-carboxamide ≥97%

Supplier: Ambeed
Warning

Synonyms: GNE-495

A165313-1MG A165313-5MG
77615-366EA 32.96 USD
77615-366 77615-368
8-Amino-N-(1-(cyclopropanecarbonyl)azetidin-3-yl)-2-(3-fluorophenyl)-1,7-naphthyridine-5-carboxamide ≥97%
8-Amino-N-(1-(cyclopropanecarbonyl)azetidin-3-yl)-2-(3-fluorophenyl)-1,7-naphthyridine-5-carboxamide

Precaución: for Research Use Only

Formula: C₂₂H₂₀FN₅O₂
MW: 405.42 g/mol
MDL Number: MFCD30489735
CAS Number: 1449277-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR