1-(2-Hydroxyethyl)-4-methyl-N-(4-(methylsulfonyl)phenyl)-5-(2-(trifluoromethyl)phenyl)-1H-pyrrole-3-carboxamide ≥98%

Supplier: Ambeed
Warning

Synonyms: Esaxerenone

A165209-1MG A165209-5MG A165209-10MG
77615-362EA 22.66 USD
77615-362 77615-364 77615-360
1-(2-Hydroxyethyl)-4-methyl-N-(4-(methylsulfonyl)phenyl)-5-(2-(trifluoromethyl)phenyl)-1H-pyrrole-3-carboxamide ≥98%
1-(2-Hydroxyethyl)-4-methyl-N-(4-(methylsulfonyl)phenyl)-5-(2-(trifluoromethyl)phenyl)-1H-pyrrole-3-carboxamide

Precaución: for Research Use Only

Formula: C₂₂H₂₁F₃N₂O₄S
MW: 466.47 g/mol
CAS Number: 1632006-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR