2-Fluoro-4-iodo-N-methylbenzamide 95+%

Supplier: Ambeed
Danger

A164939-25G A164939-5G
77892-119EA 123 USD
77892-119 77892-120
2-Fluoro-4-iodo-N-methylbenzamide 95+%
2-Fluoro-4-iodo-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₇FINO
MDL Number: MFCD26388774
CAS Number: 1351185-56-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR