6-(2-(Quinolin-2-yl)propan-2-yl)pyridin-2(1H)-one ≥98%
Supplier: Ambeed
|
Warning
|
A1648271-100MG
A1648271-250MG
A1648271-50MG
77625-622EA
154.5
USD
77625-622
77625-624
77625-626
6-(2-(Quinolin-2-yl)propan-2-yl)pyridin-2(1H)-one ≥98%
6-(2-(Quinolin-2-yl)propan-2-yl)pyridin-2(1H)-one
Precaución: for Research Use Only
Formula:
C₁₇H₁₆N₂O MW: 264.32 g/mol |
CAS Number:
2759197-62-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...