(2-(Methoxycarbonyl)pyrimidin-5-yl)boronic acid ≥96%

Supplier: Ambeed
Warning

A164800-1G A164800-250MG A164800-5G A164800-100MG
77892-116EA 514 USD
77892-116 77892-117 77892-118 77892-115
(2-(Methoxycarbonyl)pyrimidin-5-yl)boronic acid ≥96%
(2-(Methoxycarbonyl)pyrimidin-5-yl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₇BN₂O₄
MDL Number: MFCD26521712
CAS Number: 1814938-78-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR