3,5-Difluoro-N-fmoc-L-phenylalanine ≥98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Phe(3,5-DiF)-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(3,5-difluorophenyl)propanoic acid

A164379-100MG A164379-250MG A164379-1G A164379-25G A164379-5G
77600-322EA 10.25 USD
77600-322 77600-324 77667-302 77871-827 77871-828
3,5-Difluoro-N-fmoc-L-phenylalanine ≥98%
3,5-Difluoro-N-fmoc-L-phenylalanine

Precaución: for Research Use Only

Formula: C₂₄H₁₉F₂NO₄
MW: 423.4 g/mol
MDL Number: MFCD00797579
CAS Number: 205526-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR