tert-Butyl ethyl(2-oxoethyl)carbamate ≥97%

Supplier: Ambeed
Warning

A164236-1G A164236-5G
77892-110EA 177 USD
77892-110 77892-111
tert-Butyl ethyl(2-oxoethyl)carbamate ≥97%

Precaución: for Research Use Only

Formula: C₉H₁₇NO₃
MDL Number: MFCD12913662
CAS Number: 315718-06-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR