4-Chloro-6-(methoxymethyl)-2-(methylthio)pyrimidine ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A164063-100MG A164063-1G
77683-352EA 12 USD
77683-352 77683-353
4-Chloro-6-(methoxymethyl)-2-(methylthio)pyrimidine ≥95%, Ambeed.Inc
4-Chloro-6-(methoxymethyl)-2-(methylthio)pyrimidine

Precaución: For research use only.

Formula: C₇H₉ClN₂OS
MW: 204.67 g/mol
MDL Number: MFCD28401038
CAS Number: 1499503-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR