[Ir(ppy)2(4,4′-(NH2)2,5,5′-Me2bpy)]PF6 ≥95%

Supplier: Ambeed
Warning

Synonyms: (OC-6-33)-(5,5′-Dimethyl[2,2′-bipyridine]-4,4′-diamine-κN1,κN1′)bis[2-(2-pyridinyl-κN)phenyl-κC]-Iridium(1+), hexafluorophosphate(1-) (1:1)

A1638456-100MG A1638456-50MG A1638456-250MG
77592-992EA 296.23 USD
77592-992 77592-994 77876-489
[Ir(ppy)2(4,4′-(NH2)2,5,5′-Me2bpy)]PF6 ≥95%
[Ir(ppy)2(4,4′-(NH2)2,5,5′-Me2bpy)]PF6

Precaución: for Research Use Only

Formula: C₃₄H₃₀F₆IrN₆P
MW: 859.82 g/mol
CAS Number: 2489240-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR