2,5-Dimethoxy-4-formylphenylboronic acid pinacol ester ≥98%

Supplier: Ambeed
Warning

A1633900-1G A1633900-250MG A1633900-5G
77876-469EA 149 USD
77876-469 77876-470 77876-471
2,5-Dimethoxy-4-formylphenylboronic acid pinacol ester ≥98%
2,5-Dimethoxy-4-formylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₁BO₅
MDL Number: MFCD32200312
CAS Number: 2633632-66-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR