6-Hydroxy-1-methyl-1H-indazole ≥98%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-1H-indazol-6-ol

A163110-250MG A163110-100MG A163110-1G
77600-312EA 70.73 USD
77600-312 77667-276 77667-278
6-Hydroxy-1-methyl-1H-indazole ≥98%
6-Hydroxy-1-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₈N₂O
MW: 148.162 g/mol
MDL Number: MFCD00456074
CAS Number: 118933-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR