4,4,5,5-Tetramethyl-2-(4-(phenylsulfonyl)phenyl)-1,3,2-dioxaborolane ≥98%

Supplier: Ambeed
Warning

A1625851-1G A1625851-250MG
77876-440EA 262 USD
77876-440 77876-441
4,4,5,5-Tetramethyl-2-(4-(phenylsulfonyl)phenyl)-1,3,2-dioxaborolane ≥98%
4,4,5,5-Tetramethyl-2-(4-(phenylsulfonyl)phenyl)-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₈H₂₁BO₄S
MDL Number: MFCD31641436
CAS Number: 1361215-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR