Ethyl 2-formylthiazole-4-carboxylate ≥95%

Supplier: Ambeed
Warning

A162140-1G A162140-5G
77592-970EA 48.18 USD
77592-970 77667-270
Ethyl 2-formylthiazole-4-carboxylate ≥95%
Ethyl 2-formylthiazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₇H₇NO₃S
MW: 185.2 g/mol
MDL Number: MFCD08689563
CAS Number: 73956-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR