(6-Bromonaphthalen-2-yl)boronic acid ≥98%

Supplier: Ambeed
Warning

A1620613-100MG A1620613-1G A1620613-250MG
77876-425EA 48 USD
77876-425 77876-426 77876-427
(6-Bromonaphthalen-2-yl)boronic acid ≥98%
(6-Bromonaphthalen-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₈BBrO₂
MW: 250.88 g/mol
MDL Number: MFCD18410711
CAS Number: 1337916-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR