1-(3-Iodophenyl)ethan-1-ol ≥95%

Supplier: Ambeed
Warning

A1619610-1G A1619610-5G
77592-966EA 194.75 USD
77592-966 77592-968
1-(3-Iodophenyl)ethan-1-ol ≥95%
1-(3-Iodophenyl)ethan-1-ol

Precaución: for Research Use Only

Formula: C₈H₉IO
MDL Number: MFCD22573005
CAS Number: 79917-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR