((S)-2-Hydroxypropanoyl)-L-phenylalanine ≥95%
Supplier: Ambeed
|
Warning
|
A1619554-10MG
A1619554-1MG
A1619554-5MG
A1619554-100MG
A1619554-50MG
77598-120EA
105.58
USD
77598-120
77598-122
77598-124
77876-422
77876-423
((S)-2-Hydroxypropanoyl)-L-phenylalanine ≥95%
((S)-2-Hydroxypropanoyl)-L-phenylalanine
Precaución: for Research Use Only
Formula:
C₁₂H₁₅NO₄ |
MDL Number:
MFCD34676338 CAS Number: 183241-73-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...