1-(tert-Butoxycarbonyl)-3-fluoropyrrolidine-3-carboxylic acid ≥98%

Supplier: Ambeed
Warning

A161801-100MG A161801-250MG
77685-326EA 87 USD
77685-326 77892-102
1-(tert-Butoxycarbonyl)-3-fluoropyrrolidine-3-carboxylic acid ≥98%
1-(tert-Butoxycarbonyl)-3-fluoropyrrolidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₆FNO₄
MDL Number: MFCD08703174
CAS Number: 1001754-59-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR