(3-Methoxy-5-nitrophenyl)boronic acid ≥95%

Supplier: Ambeed
Warning

A1617885-100MG A1617885-1G A1617885-250MG
77876-388EA 48 USD
77876-388 77876-389 77876-390
(3-Methoxy-5-nitrophenyl)boronic acid ≥95%
(3-Methoxy-5-nitrophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₈BNO₅
MDL Number: MFCD22544495
CAS Number: 2851877-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR