4-[2-(Trimethylsilyl)ethynyl]morpholine ≥95%

Supplier: Ambeed
Warning

A161690-250MG A161690-1G A161690-5G
77592-962EA 12.3 USD
77592-962 77592-960 77748-086
4-[2-(Trimethylsilyl)ethynyl]morpholine ≥95%
4-[2-(Trimethylsilyl)ethynyl]morpholine

Precaución: for Research Use Only

Formula: C₉H₁₇NOSi
MDL Number: MFCD05865184
CAS Number: 64024-63-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR