Biotin-PEG2-Amine ≥98%

Supplier: Ambeed
Warning

Synonyms: N-(2-(2-(2-Aminoethoxy)ethoxy)ethyl)-5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamide, Biotin-DADOO, N-Biotinyl-3,6-dioxaoctane-1,8-diamine, N-[2-[2-(2-Aminoethoxy)ethoxy]ethyl]biotinamide

A161581-250MG A161581-100MG A161581-50MG A161581-1G
77600-290EA 221.4 USD
77600-290 77611-568 77671-284 77883-269
Biotin-PEG2-Amine ≥98%
Biotin-PEG2-Amine

Precaución: for Research Use Only

Formula: C₁₆H₃₀N₄O₄S
MDL Number: MFCD09264745
CAS Number: 138529-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR