tert-Butyl 6-hydroxy-2-azabicyclo[2.2.1]heptane-2-carboxylate ≥96%
Supplier: Ambeed
|
Warning
|
A161570-1G
A161570-250MG
77611-566EA
33.99
USD
77611-566
77883-268
tert-Butyl 6-hydroxy-2-azabicyclo[2.2.1]heptane-2-carboxylate ≥96%
tert-Butyl 6-hydroxy-2-azabicyclo[2.2.1]heptane-2-carboxylate
Precaución: for Research Use Only
Formula:
C₁₁H₁₉NO₃ |
MDL Number:
MFCD20278316 CAS Number: 207405-59-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...