Methyl 4-amino-6-methylnicotinate ≥95%

Supplier: Ambeed
Warning

A161371-100MG A161371-250MG
77671-280EA 116 USD
77671-280 77883-263
Methyl 4-amino-6-methylnicotinate ≥95%
Methyl 4-amino-6-methylnicotinate

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MDL Number: MFCD07374917
CAS Number: 886372-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR