(2-Methoxy-6-(trifluoromethyl)phenyl)boronic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxy-6-trifluoromethylphenylboronic acid

A161312-100MG A161312-1G A161312-250MG
77901-897EA 43 USD
77901-897 77901-413 77901-051
(2-Methoxy-6-(trifluoromethyl)phenyl)boronic acid ≥95%
(2-Methoxy-6-(trifluoromethyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₈BF₃O₃
MW: 219.95 g/mol
MDL Number: MFCD08059516
CAS Number: 1310384-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR