(E)-N-(2-aminophenyl)-3-(1-((4-(1-methyl-1H-pyrazol-4-yl)phenyl)sulfonyl)-1H-pyrrol-3-yl)acrylamide 98+%

Supplier: Ambeed
Warning

Synonyms: Domatinostat

A161236-10MG A161236-2MG A161236-5MG A161236-1MG A161236-50MG
77615-318EA 37.08 USD
77615-318 77615-320 77615-322 77746-101 77746-102
(E)-N-(2-aminophenyl)-3-(1-((4-(1-methyl-1H-pyrazol-4-yl)phenyl)sulfonyl)-1H-pyrrol-3-yl)acrylamide 98+%
(E)-N-(2-aminophenyl)-3-(1-((4-(1-methyl-1H-pyrazol-4-yl)phenyl)sulfonyl)-1H-pyrrol-3-yl)acrylamide

Precaución: for Research Use Only

Formula: C₂₃H₂₁N₅O₃S
MW: 447.51 g/mol
MDL Number: MFCD28155264
CAS Number: 910462-43-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR