Benzenesulfinic acid ≥95%

Supplier: Ambeed
Warning

A161190-100MG A161190-1G A161190-250MG
77877-065EA 21 USD
77877-065 77877-066 77877-067
Benzenesulfinic acid ≥95%
Benzenesulfinic acid

Precaución: for Research Use Only

Formula: C₆H₆O₂S
MDL Number: MFCD00800458
CAS Number: 618-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR