Calycosin-7-O-β-D-Glucoside ≥98%

Supplier: Ambeed
Warning

A161040-100MG A161040-10MG A161040-5MG
77685-321EA 524 USD
77685-321 77685-322 77685-323
Calycosin-7-O-β-D-Glucoside ≥98%
Calycosin-7-O-β-D-Glucoside

Precaución: for Research Use Only

Formula: C₂₂H₂₂O₁₀
MW: 446.4 g/mol
MDL Number: MFCD08061493
CAS Number: 20633-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR