2-[2-Ethoxy-5-[(4-ethyl-1-piperazinyl)sulfonyl]phenyl]-5-methyl-7-propyl-1H-imidazo[5,1-f][1,2,4]triazin-4-one hydrochloride ≥98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Vardenafil HCl, 2-(2-Ethoxy-5-((4-ethylpiperazin-1-yl)sulphonyl)phenyl)-5-methyl-7-propylimidazo[5,1-f][1,2,4]triazin-4(1H)-one hydrochloride
A160973-5G
A160973-1G
A160973-25G
77600-282EA
82
USD
77600-282
77600-280
77667-262
2-[2-Ethoxy-5-[(4-ethyl-1-piperazinyl)sulfonyl]phenyl]-5-methyl-7-propyl-1H-imidazo[5,1-f][1,2,4]triazin-4-one hydrochloride ≥98%
2-[2-Ethoxy-5-[(4-ethyl-1-piperazinyl)sulfonyl]phenyl]-5-methyl-7-propyl-1H-imidazo[5,1-f][1,2,4]triazin-4-one hydrochloride
Precaución: for Research Use Only
Formula:
C₂₃H₃₂N₆O₄S·ClH·3H₂O |
MDL Number:
MFCD07369690 CAS Number: 224785-91-5 UN: 2811 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...