2-(2-Ethoxyphenoxy)ethanol ≥97%

Supplier: Ambeed
Warning

A160956-1G A160956-5G
77746-098EA 20 USD
77746-098 77746-099
2-(2-Ethoxyphenoxy)ethanol ≥97%
2-(2-Ethoxyphenoxy)ethanol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₃
MDL Number: MFCD09675039
CAS Number: 3250-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR