(2S,4aS,6aR,7S,9R,11aS,11bS)-7-Hydroxy-2-(((2R,3R,4R,5R,6R)-6-(hydroxymethyl)-3-((3-methylbutanoyl)oxy)-4,5-bis(sulfooxy)tetrahydro-2H-pyran-2-yl)oxy)-11b-methyl-8-methylenedodecahydro-6a,9-methanocyclohepta[a]naphthalene-4,4(4aH)-dicarboxylic acid dipotassium salt ≥98%

Supplier: Ambeed
Warning

Synonyms: Carboxyatractyloside dipotassium

A160580-10MG A160580-50MG
77615-308EA 90.64 USD
77615-308 77783-344
(2S,4aS,6aR,7S,9R,11aS,11bS)-7-Hydroxy-2-(((2R,3R,4R,5R,6R)-6-(hydroxymethyl)-3-((3-methylbutanoyl)oxy)-4,5-bis(sulfooxy)tetrahydro-2H-pyran-2-yl)oxy)-11b-methyl-8-methylenedodecahydro-6a,9-methanocyclohepta[a]naphthalene-4,4(4aH)-dicarboxylic acid dipotassium salt ≥98%
(2S,4aS,6aR,7S,9R,11aS,11bS)-7-Hydroxy-2-(((2R,3R,4R,5R,6R)-6-(hydroxymethyl)-3-((3-methylbutanoyl)oxy)-4,5-bis(sulfooxy)tetrahydro-2H-pyran-2-yl)oxy)-11b-methyl-8-methylenedodecahydro-6a,9-methanocyclohepta[a]naphthalene-4,4(4aH)-dicarboxylic acid dipot

Precaución: for Research Use Only

Formula: C₃₁H₄₆K₂O₁₈S₂
MDL Number: MFCD00152201
CAS Number: 33286-30-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR