D-Kynurenine free base ≥98%

Supplier: Ambeed
Warning

Synonyms: β-Anthraniloyl-D-alanine, (R)-2-Amino-4-(2-aminophenyl)-4-oxobutanoic acid, D-Kynurenine

A160463-100MG A160463-50MG
77685-313EA 160 USD
77685-313 77685-314
D-Kynurenine free base ≥98%
D-Kynurenine free base

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O₃
MW: 208.214 g/mol
MDL Number: MFCD00133224
CAS Number: 13441-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR