3-((3,5-Bis(trifluoromethyl)phenyl)amino)benzo[d]isothiazole 1,1-dioxide ≥98%

Supplier: Ambeed
Warning

A1604091-10MG A1604091-1MG A1604091-5MG
77598-108EA 120.95 USD
77598-108 77598-110 77598-112
3-((3,5-Bis(trifluoromethyl)phenyl)amino)benzo[d]isothiazole 1,1-dioxide ≥98%
3-((3,5-Bis(trifluoromethyl)phenyl)amino)benzo[d]isothiazole 1,1-dioxide

Precaución: for Research Use Only

Formula: C₁₅H₈F₆N₂O₂S
CAS Number: 823830-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR