(R)-7-Chloro-2-(trans-4-(3-methoxyazetidin-1-yl)cyclohexyl)-2,4-dimethyl-N-((6-methyl-4-(methylthio)-2-oxo-1,2-dihydropyridin-3-yl)methyl)benzo[d][1,3]dioxole-5-carboxamide ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Tulmimetostat
A1603652-10MG
A1603652-5MG
77625-548EA
119.48
USD
77625-548
77625-550
(R)-7-Chloro-2-(trans-4-(3-methoxyazetidin-1-yl)cyclohexyl)-2,4-dimethyl-N-((6-methyl-4-(methylthio)-2-oxo-1,2-dihydropyridin-3-yl)methyl)benzo[d][1,3]dioxole-5-carboxamide ≥98%
(R)-7-Chloro-2-(trans-4-(3-methoxyazetidin-1-yl)cyclohexyl)-2,4-dimethyl-N-((6-methyl-4-(methylthio)-2-oxo-1,2-dihydropyridin-3-yl)methyl)benzo[d][1,3]dioxole-5-carboxamide
Precaución: for Research Use Only
Formula:
C₂₈H₃₆ClN₃O₅S MW: 562.12 g/mol |
CAS Number:
2567686-02-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...