N-((S)-1-((4-(6-((2R,6S)-2,6-Dimethylmorpholino)pyridin-2-yl)thiazol-2-yl)amino)-3-methoxy-1-oxopropan-2-yl)-1-(methylsulfonyl)-1H-pyrrole-3-carboxamide 98% 99.9+% ee

Supplier: Ambeed
Warning

Synonyms: FHD-286

A1603632-1MG A1603632-5MG A1603632-10MG
77598-104EA 210.13 USD
77598-104 77598-106 77598-102
N-((S)-1-((4-(6-((2R,6S)-2,6-Dimethylmorpholino)pyridin-2-yl)thiazol-2-yl)amino)-3-methoxy-1-oxopropan-2-yl)-1-(methylsulfonyl)-1H-pyrrole-3-carboxamide 98% 99.9+% ee
N-((S)-1-((4-(6-((2R,6S)-2,6-Dimethylmorpholino)pyridin-2-yl)thiazol-2-yl)amino)-3-methoxy-1-oxopropan-2-yl)-1-(methylsulfonyl)-1H-pyrrole-3-carboxamide

Precaución: for Research Use Only

Formula: C₂₄H₃₀N₆O₆S₂
MDL Number: MFCD34662947
CAS Number: 2671128-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR