6-Fluoro-5-(4-((5-fluoro-2-methyl-3-oxo-3,4-dihydroquinoxalin-6-yl)methyl)piperazin-1-yl)-N-methylpicolinamide ≥98%

Supplier: Ambeed
Warning

Synonyms: AZD-9574

A1601840-10MG A1601840-1MG A1601840-5MG
77625-540EA 432.6 USD
77625-540 77625-542 77625-544
6-Fluoro-5-(4-((5-fluoro-2-methyl-3-oxo-3,4-dihydroquinoxalin-6-yl)methyl)piperazin-1-yl)-N-methylpicolinamide ≥98%
6-Fluoro-5-(4-((5-fluoro-2-methyl-3-oxo-3,4-dihydroquinoxalin-6-yl)methyl)piperazin-1-yl)-N-methylpicolinamide

Precaución: for Research Use Only

Formula: C₂₁H₂₂F₂N₆O₂
MW: 428.44 g/mol
CAS Number: 2756333-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR